missing translation for 'onlineSavingsMsg'
Learn More

2-chloroaniline, 98+%, Thermo Scientific Chemicals

Catalog Number p-3777502
19.46 EUR valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
250 mL
1 L
5 L
Packaging:
Glass bottle
Plastic drum
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
95-51-2
C6H6ClN
127.57
MFCD00007656
AKCRQHGQIJBRMN-UHFFFAOYSA-N
o-chloroaniline, benzenamine, 2-chloro, 2-chlorobenzenamine, 2-chlorophenylamine, 1-amino-2-chlorobenzene, o-aminochlorobenzene, o-chloroaminobenzene, aniline, o-chloro, o-chloraniline, fast yellow gc base
7240
2-chloroaniline
C1=CC=C(C(=C1)N)Cl
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
95-51-2
C6H6ClN
127.57
MFCD00007656
AKCRQHGQIJBRMN-UHFFFAOYSA-N
o-chloroaniline, benzenamine, 2-chloro, 2-chlorobenzenamine, 2-chlorophenylamine, 1-amino-2-chlorobenzene, o-aminochlorobenzene, o-chloroaminobenzene, aniline, o-chloro, o-chloraniline, fast yellow gc base
7240
2-chloroaniline
C1=CC=C(C(=C1)N)Cl

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point -3°C
CAS Min % 98.0
Color Yellow to Brown
CAS Max % 100.0
Density 1.2100g/mL
Boiling Point 209°C
Quantity 250 mL
Infrared Spectrum Authentic
Packaging Glass bottle
Physical Form Liquid
Refractive Index 1.5870 to 1.589
Assay Percent Range 98% min. (GC)
Specific Gravity 1.21
Linear Formula ClC6H4NH2
Beilstein 12, 597
Merck Index 15, 2118
Solubility Information Solubility in water: 5.13g/L (20°C). Other solubilities: soluble in most common organic solvents,miscible with alcohol and ether
Formula Weight 127.57
Percent Purity 98+%
Chemical Name or Material 2-Chloroaniline
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 2-Chloroaniline
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H373-May cause damage to organs.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'