missing translation for 'onlineSavingsMsg'
Learn More

2-Chlorobenzaldehyde, 97%

Product Code. p-7021234
Click to view available options
Quantity:
10 g
250 g
1000 g
Unit Size:
1000g
10g
250g
This item is not returnable. View return policy

Product Code. 11319649

Brand: Thermo Scientific Alfa Aesar A13488.09

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 89-98-5 | C7H5ClO | 140.566 g/mol

2-Chlorobenzaldehyde has been used in generation of small focused library of diversely functionalized dihydropyrimidine derivatives via one-pot three-component Biginelli cyclocondensation of β-ketoesters, aldehydes and thioureas

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Chlorobenzaldehyde has been used in generation of small focused library of diversely functionalized dihydropyrimidine derivatives via one-pot three-component Biginelli cyclocondensation of β-ketoesters, aldehydes and thioureas

Solubility
Soluble in water (1.8g/L), alcohol, ether, benzene, and acetone.

Notes
Air Sensitive. Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 89-98-5
Molecular Formula C7H5ClO
Molecular Weight (g/mol) 140.566
MDL Number MFCD00003304
InChI Key FPYUJUBAXZAQNL-UHFFFAOYSA-N
Synonym o-chlorobenzaldehyde, chlorobenzaldehyde, benzaldehyde, 2-chloro, benzaldehyde, o-chloro, 2-chlorbenzaldehyd, 2-clorobenzaldeide, o-chloorbenzaldehyde, usaf m-7, 2-chloorbenzaldehyde, o-chlorobenzenecarboxaldehyde
PubChem CID 6996
IUPAC Name 2-chlorobenzaldehyde
SMILES C1=CC=C(C(=C1)C=O)Cl

Specifications

Melting Point 10°C to 12°C
Density 1.249
pH 2.9
Boiling Point 212°C
Flash Point 87°C (188°F)
Odor Pungent
Refractive Index 1.5675
Quantity 10 g
UN Number UN3265
Beilstein 385877
Sensitivity Air Sensitive
Solubility Information Soluble in water (1.8g/L),alcohol,ether,benzene,and acetone.
Formula Weight 140.57
Percent Purity 97%
Chemical Name or Material 2-Chlorobenzaldehyde
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.