missing translation for 'onlineSavingsMsg'
Learn More

2-Chlorobenzimidazole, 97%

Product Code. p-7081927
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11404853

Brand: Thermo Scientific Alfa Aesar A12193.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 4857-06-1 | C7H5ClN2 | 152.58 g/mol

2-Chlorobenzimidazole is mainly used for the synthesis of medicine and as an intermediate in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Chlorobenzimidazole is mainly used for the synthesis of medicine and as an intermediate in organic synthesis.

Solubility
Insoluble in water.

Notes
Store in cool, dry place, in well sealed container. Store away from oxidizing agents, alkali metals, bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4857-06-1
MDL Number MFCD00051944

Specifications

Melting Point ∼180°C to 185°C
Molecular Formula C16H33Cl
Quantity 5 g
Beilstein 116526
Synonym cetyl chloride, hexadecyl chloride, hexadecane, 1-chloro, n-hexadecyl chloride, chlorohexadecane, unii-wq5zks3ek3, wq5zks3ek3, 1-chloro-hexadecane, 1-chloranylhexadecane, dsstox_cid_7580
Solubility Information Insoluble in water.
InChI Key CLWAXFZCVYJLLM-UHFFFAOYSA-N
SMILES CCCCCCCCCCCCCCCCCl
IUPAC Name 1-chlorohexadecane
Molecular Weight (g/mol) 260.89
PubChem CID 20993
Formula Weight 152.58
Percent Purity 97%
Chemical Name or Material 2-Chlorobenzimidazole
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.