missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Chlorobenzyl mercaptan, 98%, Thermo Scientific™
Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
Specifications
Specifications
| CAS Min % | 97.5 |
| Color | Colorless to Yellow |
| CAS Max % | 100.0 |
| Density | 1.2200g/mL |
| Boiling Point | 120°C to 121°C (25.0 mmHg) |
| Quantity | 25 g |
| Infrared Spectrum | Authentic |
| Packaging | Glass bottle |
| Physical Form | Liquid |
| Refractive Index | 1.5890 to 1.591 |
| Show More |
Safety and Handling
Product Identifier
- 2-Chlorobenzyl mercaptan
- Warning
- Acute toxicity Category 4
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- H302-Harmful if swallowed.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction