missing translation for 'onlineSavingsMsg'
Learn More

2-Chloroethyl methacrylate, 97%, stab. with 400-600ppm hydroquinone

Product Code. 11352267
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
This item is not returnable. View return policy

Product Code. 11352267

Brand: Thermo Scientific Alfa Aesar L09783.09

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1888-94-4 | C6H9ClO2 | 148.59 g/mol

2-Chloroethyl methacrylate is used as a monomer to prepare poly(2-chloroethyl methacrylate).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Chloroethyl methacrylate is used as a monomer to prepare poly(2-chloroethyl methacrylate).

Solubility
Immiscible with water.

Notes
Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1888-94-4
Molecular Formula C6H9ClO2
Molecular Weight (g/mol) 148.59
MDL Number MFCD00045291
InChI Key GPOGMJLHWQHEGF-UHFFFAOYSA-N
Synonym 2-chloroethyl methacrylate, chloroethyl methacrylate, methacrylic acid, 2-chloroethyl ester, 2-propenoic acid, 2-methyl-, 2-chloroethyl ester, .beta.-chloroethyl methacrylate, methacrylic acid beta-chloroethyl ester, methacrylic acid .beta.-chloroethyl ester, acmc-20anxx, chloroethylmethacrylate, 2-chloroethylmethacrylate
PubChem CID 15905
IUPAC Name 2-chloroethyl 2-methylprop-2-enoate
SMILES CC(=C)C(=O)OCCCl

Specifications

Boiling Point 170°C
Flash Point 69°C (156°F)
Refractive Index 1.451
Quantity 10 g
UN Number UN2810
Beilstein 956599
Solubility Information Immiscible with water.
Formula Weight 148.59
Percent Purity 97%
Chemical Name or Material 2-Chloroethyl methacrylate, stabilized with 500ppm hydroquinone

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.