missing translation for 'onlineSavingsMsg'
Learn More

2-chloroethylamine hydrochloride, 98%, Thermo Scientific Chemicals

Catalog Number p-3757225
Click to view available options
Quantity:
25 g
100 g
1 kg
Packaging:
Glass bottle
Plastic bottle
This item is not returnable. View return policy
870-24-6
C2H7Cl2N
115.99
MFCD00012887,MFCD00137399
ONRREFWJTRBDRA-UHFFFAOYSA-N
2-chloroethylamine hydrochloride, 2-chloroethanamine hydrochloride, 1-amino-2-chloroethane hydrochloride, ethanamine, 2-chloro-, hydrochloride, 2-aminoethyl chloride hydrochloride, ethanamine, 2-chloro-, hydrochloride 1:1, 2-chloroethan-1-amine hydrochloride, chlorethamine hydrochloride, 2-chloro-ethylamine hydrochloride, chlorethamine hcl
9793737
[Cl-].[NH3+]CCCl
This item is not returnable. View return policy
870-24-6
C2H7Cl2N
115.99
MFCD00012887,MFCD00137399
ONRREFWJTRBDRA-UHFFFAOYSA-N
2-chloroethylamine hydrochloride, 2-chloroethanamine hydrochloride, 1-amino-2-chloroethane hydrochloride, ethanamine, 2-chloro-, hydrochloride, 2-aminoethyl chloride hydrochloride, ethanamine, 2-chloro-, hydrochloride 1:1, 2-chloroethan-1-amine hydrochloride, chlorethamine hydrochloride, 2-chloro-ethylamine hydrochloride, chlorethamine hcl
9793737
[Cl-].[NH3+]CCCl

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 140.0°C to 150.0°C
Color White to Beige
Loss on Drying 1% max.
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula ClCH2CH2NH2·HCl
Quantity 25 g
Beilstein 04,133
Solubility Information Solubility in water: soluble in water
IUPAC Name 2-chloroethan-1-aminium chloride
Formula Weight 115.99
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 2-Chloroethylamine hydrochloride
Show More Show Less
compliance-icons
Product Identifier
  • 2-Chloroethylamine hydrochloride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'