missing translation for 'onlineSavingsMsg'
Learn More

2-chloromalonaldehyde, 95%, Thermo Scientific Chemicals

Catalog Number p-3735981
Click to view available options
Quantity:
1 g
10 g
50 g
Packaging:
Glass bottle
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
36437-19-1
MFCD01320489
KTRZQCIGJUWSGE-UHFFFAOYSA-N
2-chloromalonaldehyde, chloromalonaldehyde, propanedial, chloro, chloromalonalde-hyde, 2-chloro-malonaldehyde, propanedial, 2-chloro, 2-chloropropane-1,3-dial, chloromalonaldehyde; chloromalondialdehyde; chloromalonic dialdehyde;2-chloromalonaldehyde
147646
2-chloropropanedial
C(=O)C(C=O)Cl
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
36437-19-1
MFCD01320489
KTRZQCIGJUWSGE-UHFFFAOYSA-N
2-chloromalonaldehyde, chloromalonaldehyde, propanedial, chloro, chloromalonalde-hyde, 2-chloro-malonaldehyde, propanedial, 2-chloro, 2-chloropropane-1,3-dial, chloromalonaldehyde; chloromalondialdehyde; chloromalonic dialdehyde;2-chloromalonaldehyde
147646
2-chloropropanedial
C(=O)C(C=O)Cl

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 94.0
CAS Max % 100.0
Melting Point 140.0°C to 145.0°C
Color Brown
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass bottle
Molecular Formula C3H3ClO2
Linear Formula ClCH(CHO)2
Quantity 1 g
Solubility Information Solubility in water: soluble. Other solubilities: soluble in chloroform,benzene,dimethylsulfoxide
Molecular Weight (g/mol) 106.5
Formula Weight 106.5
Percent Purity 95%
Physical Form Crystalline Powder
Chemical Name or Material 2-chloromalonaldehyde
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'