missing translation for 'onlineSavingsMsg'
Learn More

2-Chloropyridine N-oxide hydrochloride, 97%, Thermo Scientific Chemicals

Catalog Number 11322329
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
20295-64-1
C5H5Cl2NO
166.001
MFCD00007913
GRZNODNSNCXOHE-UHFFFAOYSA-N
2-chloropyridine n-oxide hydrochloride, 2-chloropyridine-n-oxide hydrochloride, 2-chloropyridine 1-oxide hydrochloride, chloride 2-chloropyridinium 1-oxide, 2-chloropyridine n-oxide hcl, pyridine, 2-chloro-, 1-oxide, hydrochloride, 2-chloropyridin-1-ium-1-olate hydrochloride, 2-chlorpyridin-1-oxidhydrochlorid, acmc-1ccui, 2-chloropyridine n-oxide.hcl
88478
2-chloro-1-oxidopyridin-1-ium;hydrochloride
C1=CC=[N+](C(=C1)Cl)[O-].Cl
This item is not returnable. View return policy
20295-64-1
C5H5Cl2NO
166.001
MFCD00007913
GRZNODNSNCXOHE-UHFFFAOYSA-N
2-chloropyridine n-oxide hydrochloride, 2-chloropyridine-n-oxide hydrochloride, 2-chloropyridine 1-oxide hydrochloride, chloride 2-chloropyridinium 1-oxide, 2-chloropyridine n-oxide hcl, pyridine, 2-chloro-, 1-oxide, hydrochloride, 2-chloropyridin-1-ium-1-olate hydrochloride, 2-chlorpyridin-1-oxidhydrochlorid, acmc-1ccui, 2-chloropyridine n-oxide.hcl
88478
2-chloro-1-oxidopyridin-1-ium;hydrochloride
C1=CC=[N+](C(=C1)Cl)[O-].Cl

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 138°C to 142°C
Quantity 5 g
Formula Weight 166.01
Percent Purity 97%
Chemical Name or Material 2-Chloropyridine N-oxide hydrochloride

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'