missing translation for 'onlineSavingsMsg'
Learn More

2-Chloropyrimidine, 98+%, Thermo Scientific Chemicals

Catalog Number p-4509372
Click to view available options
Quantity:
10 g
50 g
250 g
Packaging:
Glass bottle
This item is not returnable. View return policy
1722-12-9
C4H3ClN2
114.53
MFCD00006060
UNCQVRBWJWWJBF-UHFFFAOYSA-N
pyrimidine, 2-chloro, 2-chloro-pyrimidine, chloropyrimidine, 2-chloro pyrimidine, 2-chloro-1,3-diazine, 2-chlorpyrimidin, 2-chloro-pyrimindine, 2chloropyrimidine, chloro-pyrimidine, 2-chloropyrimidin
74404
2-chloropyrimidine
ClC1=NC=CC=N1
This item is not returnable. View return policy
1722-12-9
C4H3ClN2
114.53
MFCD00006060
UNCQVRBWJWWJBF-UHFFFAOYSA-N
pyrimidine, 2-chloro, 2-chloro-pyrimidine, chloropyrimidine, 2-chloro pyrimidine, 2-chloro-1,3-diazine, 2-chlorpyrimidin, 2-chloro-pyrimindine, 2chloropyrimidine, chloro-pyrimidine, 2-chloropyrimidin
74404
2-chloropyrimidine
ClC1=NC=CC=N1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 62.0°C to 66.0°C
Color Orange to Yellow
Boiling Point 75.0°C to 76.0°C (10.0 mmHg)
Flash Point 98°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Quantity 10 g
Formula Weight 114.53
Percent Purity 98+%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material 2-Chloropyrimidine
Show More Show Less
compliance-icons
Product Identifier
  • 2-Chloropyrimidine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'