Learn More
Julabo™ 2-Collar Nuts
Product Code. 15214333
Click to view available options
Type:
M24 x 1.5 Connections
M30 x 1.5 Connections
M38 x 1.5 Connections
Unit Size:
Pack of 2
Product Code. 15214333
Brand: Julabo™ 8970495
Only null left
Add to basket
Add to basket
Specifications
Description 2-Collar Nuts
For Use With (Equipment) Suitable for PRESTO A30, PRESTO A40, PRESTO A45, PRESTO A45t, PRESTO A80 Temperature Control System
Type M24 x 1.5 Connections