missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-(Difluoromethoxy)benzyl bromide, 97%

Brand:  Alfa Aesar™ B20931.09

Additional Details : CAS : 85684-64-6 Weight : 0.01000kg Transport : UN number : 3265 Chem class : 8 Pack group : II

Quantity 10g

 View more versions of this product

Product Code. 11489976

  • 192.00€ / 10g


Estimated Shipment
27-05-2021
Add to basket

Description

Description

Applications
2-(Difluoromethoxy)benzyl bromide is used as a building block in organic synthesis. It also serves as a coupling reagent and N-protecting reagent. Further, it is used as a gas chromatography derivatizing agent.

Solubility
Immiscible with water.

Notes
Incompatible with oxidizing agents, strong bases, alcohols and ammonia.
Specifications

Chemical Identifiers

85684-64-6
237.044
JSYYKHKAGPBIHF-UHFFFAOYSA-N
2736998
C1=CC=C(C(=C1)CBr)OC(F)F
C8H7BrF2O
MFCD00190103
1-bromomethyl-2-difluoromethoxy benzene, 2-difluoromethoxy benzyl bromide, o-difluoromethoxybenzylbromide, alpha-bromo-2-difluoromethoxytoluene, benzene, 1-bromomethyl-2-difluoromethoxy, 2-bromomethyl phenoxy difluoromethane, ksc497q8n, 2-difluoromethoxybenzyl bromide, 2-difluoromethoxy benzylbromide, 2-difluoromethoxy-benzyl bromide
1-(bromomethyl)-2-(difluoromethoxy)benzene

Specifications

2-(Difluoromethoxy)benzyl bromide
108°C (228°F)
222°C
85684-64-6
MFCD00190103
1-bromomethyl-2-difluoromethoxy benzene, 2-difluoromethoxy benzyl bromide, o-difluoromethoxybenzylbromide, alpha-bromo-2-difluoromethoxytoluene, benzene, 1-bromomethyl-2-difluoromethoxy, 2-bromomethyl phenoxy difluoromethane, ksc497q8n, 2-difluoromethoxybenzyl bromide, 2-difluoromethoxy benzylbromide, 2-difluoromethoxy-benzyl bromide
JSYYKHKAGPBIHF-UHFFFAOYSA-N
1-(bromomethyl)-2-(difluoromethoxy)benzene
2736998
97%
1.555
1.518
10g
C8H7BrF2O
UN3265
Immiscible with water.
C1=CC=C(C(=C1)CBr)OC(F)F
237.044
237.04
Videos
SDS Documents