missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(Difluoromethoxy)benzyl bromide, 97%, Thermo Scientific™
Click to view available options
Quantity:
10 g
50 g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
2-(Difluoromethoxy)benzyl bromide is used as a building block in organic synthesis. It also serves as a coupling reagent and N-protecting reagent. Further, it is used as a gas chromatography derivatizing agent.
Solubility
Immiscible with water.
Notes
Incompatible with oxidizing agents, strong bases, alcohols and ammonia.
2-(Difluoromethoxy)benzyl bromide is used as a building block in organic synthesis. It also serves as a coupling reagent and N-protecting reagent. Further, it is used as a gas chromatography derivatizing agent.
Solubility
Immiscible with water.
Notes
Incompatible with oxidizing agents, strong bases, alcohols and ammonia.
Specifications
Density | 1.555 |
Boiling Point | 222°C |
Flash Point | 108°C (228°F) |
Refractive Index | 1.518 |
Quantity | 10 g |
UN Number | UN3265 |
Solubility Information | Immiscible with water. |
Formula Weight | 237.04 |
Percent Purity | 97% |
Chemical Name or Material | 2-(Difluoromethoxy)benzyl bromide |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'