missing translation for 'onlineSavingsMsg'
Learn More

2-Difluoromethyl-1-fluorobenzene, 97%, Thermo Scientific™

Catalog Number 15406547
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
63878-70-6
C7H5F3
146.112
MFCD06657985
DBXNELKUWAMRKY-UHFFFAOYSA-N
1-difluoromethyl-2-fluorobenzene, 2-difluoromethyl-1-fluorobenzene, benzene, 1-difluoromethyl-2-fluoro, benzene, 1-difluoromethyl-2-fluoro-9ci
2761205
1-(difluoromethyl)-2-fluorobenzene
C1=CC=C(C(=C1)C(F)F)F
This item is not returnable. View return policy
63878-70-6
C7H5F3
146.112
MFCD06657985
DBXNELKUWAMRKY-UHFFFAOYSA-N
1-difluoromethyl-2-fluorobenzene, 2-difluoromethyl-1-fluorobenzene, benzene, 1-difluoromethyl-2-fluoro, benzene, 1-difluoromethyl-2-fluoro-9ci
2761205
1-(difluoromethyl)-2-fluorobenzene
C1=CC=C(C(=C1)C(F)F)F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Boiling Point 72°C to 74°C (150 mmHg)
Quantity 5 g
Formula Weight 146.11
Percent Purity 97%
Chemical Name or Material 2-Difluoromethyl-1-fluorobenzene
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'