missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(Dimethylamino)ethanethiol Hydrochloride, TRC
Click to view available options
Quantity:
2.5 g
Unit Size:
2.5g
Specifications
Specifications
| Chemical Name or Material | 2-(Dimethylamino)ethanethiol Hydrochloride |
| CAS | 13242-44-9 |
| Molecular Formula | C4H12ClNS |
| Quantity | 2.5 g |
| Synonym | (2-Mercaptoethyl)dimethylammonium Chloride, 1-(Dimethylamino)-2-mercaptoethane Hydrochloride, 2-(Dimethylamino)ethanethiol Hydrochloride, 2-(Dimethylamino)ethyl Mercaptan Hydrochloride, 2-(N,N-Dimethylamino)ethanethiol Hydrochloride Captamine Hydrochloride, MEDA, N,N-Dimethyl-2-aminoethanethiol Hydrochloride, N,N-Dimethylcysteamine Hydrochloride, N-(2-Mercaptoethyl)dimethylamine Hydrochloride, NSC 45463, Thiofluor |
| InChI Formula | InChI=1S/C4H11NS.ClH/c1-5(2)3-4-6;/h6H,3-4H2,1-2H3;1H |
| SMILES | SCCN(C)C.[H]Cl |
| IUPAC Name | 2-(dimethylamino)ethanethiol;hydrochloride |
| Molecular Weight (g/mol) | 141.66 |
| Formula Weight | 141.0379 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction