missing translation for 'onlineSavingsMsg'
Learn More

2-Ethoxy-5-pyridineboronic acid, 97%, Thermo Scientific™

Catalog Number 15284328
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
612845-44-0
C7H10BNO3
166.97
UONCERAQKBPLML-UHFFFAOYSA-N
6-ethoxypyridin-3-yl boronic acid, 2-ethoxy-5-pyridineboronic acid, 2-ethoxypyridine-5-boronic acid, 6-ethoxypyridine-3-boronic acid, 6-ethoxy-3-pyridinylboronic acid, 2-ethoxy-5-pyridylboronic acid, 3-borono-6-ethoxypyridine, 6-ethoxy-3-pyridylboronic acid, boronic acid, 6-ethoxy-3-pyridinyl, 6-ethoxy-3-pyridyl boronic acid
12012237
(6-ethoxypyridin-3-yl)boronic acid
B(C1=CN=C(C=C1)OCC)(O)O
This item is not returnable. View return policy
612845-44-0
C7H10BNO3
166.97
UONCERAQKBPLML-UHFFFAOYSA-N
6-ethoxypyridin-3-yl boronic acid, 2-ethoxy-5-pyridineboronic acid, 2-ethoxypyridine-5-boronic acid, 6-ethoxypyridine-3-boronic acid, 6-ethoxy-3-pyridinylboronic acid, 2-ethoxy-5-pyridylboronic acid, 3-borono-6-ethoxypyridine, 6-ethoxy-3-pyridylboronic acid, boronic acid, 6-ethoxy-3-pyridinyl, 6-ethoxy-3-pyridyl boronic acid
12012237
(6-ethoxypyridin-3-yl)boronic acid
B(C1=CN=C(C=C1)OCC)(O)O
Melting Point 167.0°C to 168.0°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Quantity 5 g
Formula Weight 166.97
Percent Purity 97%
Chemical Name or Material 2-Ethoxy-5-pyridineboronic acid
compliance-icons
Product Identifier
  • 2-Ethoxy-5-pyridineboronic acid, 97%
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'