missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Ethoxypyridine, TRC
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Specifications
Specifications
| Chemical Name or Material | 2-Ethoxypyridine |
| CAS | 14529-53-4 |
| Molecular Formula | C7H9NO |
| Quantity | 100 mg |
| InChI Formula | InChI=1S/C7H9NO/c1-2-9-7-5-3-4-6-8-7/h3-6H,2H2,1H3 |
| SMILES | CCOc1ccccn1 |
| IUPAC Name | 2-ethoxypyridine |
| Molecular Weight (g/mol) | 123.15 |
| Formula Weight | 123.0684 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction