missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-5-methoxybenzoic acid, 97+%, Thermo Scientific Chemicals

Catalog Number 15409047
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
367-83-9
C8H7FO3
170.139
MFCD00272566
REDSLTKMNCCQBC-UHFFFAOYSA-N
2774543
2-fluoro-5-methoxybenzoic acid
COC1=CC(=C(C=C1)F)C(=O)O
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
367-83-9
C8H7FO3
170.139
MFCD00272566
REDSLTKMNCCQBC-UHFFFAOYSA-N
2774543
2-fluoro-5-methoxybenzoic acid
COC1=CC(=C(C=C1)F)C(=O)O

2-Fluoro-5-methoxybenzoic acid is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Fluoro-5-methoxybenzoic acid is used as an organic chemical synthesis intermediate.

Notes
Stable under recommended storage conditions. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY
Quantity 1 g
Synonym 2-fluoro-5-methoxy-benzoic acid, benzoic acid,2-fluoro-5-methoxy, benzoic acid, 2-fluoro-5-methoxy, pubchem4966, 6-fluoro-m-anisic acid, 3-carboxy-4-fluoroanisole, 2-fluoro-5-methoxybenzoicacid, 6-fluoro-3-methoxybenzoic acid, rarechem al bo 0748, buttpark 45\01-62
Formula Weight 170.14
Percent Purity ≥97%
Chemical Name or Material 2-Fluoro-5-methoxybenzoic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'