missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Fluoro-5-methoxybenzoic acid, 97+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 mg
1 g
5 g
Description
2-Fluoro-5-methoxybenzoic acid is used as an organic chemical synthesis intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Fluoro-5-methoxybenzoic acid is used as an organic chemical synthesis intermediate.
Notes
Stable under recommended storage conditions. Store away from oxidizing agents.
2-Fluoro-5-methoxybenzoic acid is used as an organic chemical synthesis intermediate.
Notes
Stable under recommended storage conditions. Store away from oxidizing agents.
Specifications
Quantity | 250 mg |
Synonym | 2-fluoro-5-methoxy-benzoic acid, benzoic acid,2-fluoro-5-methoxy, benzoic acid, 2-fluoro-5-methoxy, pubchem4966, 6-fluoro-m-anisic acid, 3-carboxy-4-fluoroanisole, 2-fluoro-5-methoxybenzoicacid, 6-fluoro-3-methoxybenzoic acid, rarechem al bo 0748, buttpark 45\01-62 |
Formula Weight | 170.14 |
Percent Purity | ≥97% |
Chemical Name or Material | 2-Fluoro-5-methoxybenzoic acid |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'