missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-6-nitrobenzyl bromide, 98%, Thermo Scientific™

Catalog Number 13571610
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
1958-93-6
C7H5BrFNO2
234.02
MFCD00042918
KKSODTKRSQTJFZ-UHFFFAOYSA-N
74779
2-(bromomethyl)-1-fluoro-3-nitrobenzene
[O-][N+](=O)C1=CC=CC(F)=C1CBr
This item is not returnable. View return policy
1958-93-6
C7H5BrFNO2
234.02
MFCD00042918
KKSODTKRSQTJFZ-UHFFFAOYSA-N
74779
2-(bromomethyl)-1-fluoro-3-nitrobenzene
[O-][N+](=O)C1=CC=CC(F)=C1CBr
Melting Point 50°C to 54°C
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula O2NC6H3CH2Br(F)
Quantity 1 g
Synonym 2-fluoro-6-nitrobenzyl bromide, 2-bromomethyl-1-fluoro-3-nitrobenzene, benzene, 2-bromomethyl-1-fluoro-3-nitro, alpha-bromo-2-fluoro-6-nitrotoluene, acmc-20anuk, 2-fluoro-6-nitrobenzylbromide, 2-bromomethyl-3-fluoro-1-nitrobenzene, benzene,2-bromomethyl-1-fluoro-3-nitro
Solubility Information Solubility in water: insoluble
Formula Weight 234.02
Percent Purity 98%
Chemical Name or Material 2-Fluoro-6-nitrobenzyl bromide
compliance-icons
Product Identifier
  • 2-Fluoro-6-nitrobenzyl bromide
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'