missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Fluoroethanol, 95%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | -26.5°C |
Density | 1.1080g/mL |
Boiling Point | 100.0°C to 104.0°C |
Flash Point | 34°C |
Assay Percent Range | 94% min. (GC) |
Packaging | Glass bottle |
Linear Formula | FCH2CH2OH |
Quantity | 5 g |
Beilstein | 01,I,17 |
Specific Gravity | 1.108 |
Show More |
Safety and Handling
Product Identifier
- 2-Fluoroethanol
- Danger
- Acute toxicity Category 1
- Flammable liquid Category 3
- H300-Fatal if swallowed.
- H310-Fatal in contact with skin.
- H330-Fatal if inhaled.
- H226-Flammable liquid and vapour.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'