missing translation for 'onlineSavingsMsg'
Learn More

2-Furyltrimethysilane, 95%, Thermo Scientific™

Product Code. 15525966 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 15525966

Brand: Thermo Scientific Acros 457370010

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 1578-33-2
Molecular Formula C7H12OSi
Molecular Weight (g/mol) 140.26
InChI Key HRWRZEKRKZRDRW-UHFFFAOYSA-N
Synonym 2-trimethylsilylfuran, 2-furyltrimethylsilane, 2-furyl trimethyl silane, 5-trimethylsilylfuran, 2-trimethylsilyl furan, trimethyl 2-furyl silane, 2-trimethylsilyl-furan, amtsi020, furan-2-yl trimethyl silane
PubChem CID 137092
IUPAC Name furan-2-yl(trimethyl)silane
SMILES C[Si](C)(C)C1=CC=CO1

Specifications

Boiling Point 58.0°C (20.0 mmHg)
Flash Point 35°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 140.26
Percent Purity 95%
Chemical Name or Material 2-Furyltrimethysilane
compliance-icons compliance-icons
Product Identifier
  • 2-Furyltrimethysilane, 95%
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.