missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Hydroxy-5-iodobenzoic acid, TRC
Click to view available options
Quantity:
25 g
Unit Size:
25g
Specifications
Specifications
| Chemical Name or Material | rac 3-Hydroxy-5-keto Fluvastatin (Fluvastatin Impurity) |
| CAS | 119-30-2 |
| Molecular Formula | C7 H5 I O3 |
| Quantity | 25 g |
| InChI Formula | InChI=1S/C7H5IO3/c8-4-1-2-6(9)5(3-4)7(10)11/h1-3,9H,(H,10,11) |
| SMILES | OC(=O)c1cc(I)ccc1O |
| IUPAC Name | 2-hydroxy-5-iodobenzoic acid |
| Molecular Weight (g/mol) | 264.02 |
| Formula Weight | 263.9283 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction