missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxybenzothiazole, 98%, Thermo Scientific Chemicals

Catalog Number p-7023264
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
934-34-9
C7H5NOS
151.183
MFCD00022868
YEDUAINPPJYDJZ-UHFFFAOYSA-N
benzothiazolone, 2-hydroxybenzothiazole, 2 3h-benzothiazolone, 2-benzothiazolol, 2-benzothiazolone, 1,3-benzothiazol-2-ol, 2-benzothiazolinone, 3h-benzothiazol-2-one, benzothiazolol, benzo d thiazol-2 3h-one
13625
CHEBI:115196
3H-1,3-benzothiazol-2-one
C1=CC=C2C(=C1)NC(=O)S2
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
934-34-9
C7H5NOS
151.183
MFCD00022868
YEDUAINPPJYDJZ-UHFFFAOYSA-N
benzothiazolone, 2-hydroxybenzothiazole, 2 3h-benzothiazolone, 2-benzothiazolol, 2-benzothiazolone, 1,3-benzothiazol-2-ol, 2-benzothiazolinone, 3h-benzothiazol-2-one, benzothiazolol, benzo d thiazol-2 3h-one
13625
CHEBI:115196
3H-1,3-benzothiazol-2-one
C1=CC=C2C(=C1)NC(=O)S2

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 136°C to 140°C
Quantity 5 g
Formula Weight 151.18
Percent Purity 98%
Chemical Name or Material 2-Hydroxybenzothiazole

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'