missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxypyridine-4-carboxylic acid, 98%, Thermo Scientific Chemicals

Catalog Number p-7055617
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
22282-72-0
C6H5NO3
139.11
MFCD01318566
BXHCJLRTXPHUGH-UHFFFAOYSA-N
2-hydroxyisonicotinic acid, 2-hydroxy-4-pyridinecarboxylic acid, 2-oxo-1,2-dihydropyridine-4-carboxylic acid, 2-hydroxypyridine-4-carboxylic acid, 2-hydroxy-isonicotinic acid, 2-hydroxyisonicotinicacid, 4-pyridinecarboxylic acid, 1,2-dihydro-2-oxo, pubchem16780, acmc-209yjf, 2-hydroxy isonicotinic acid
280756
OC(=O)C1=CC(=O)NC=C1
This item is not returnable. View return policy
22282-72-0
C6H5NO3
139.11
MFCD01318566
BXHCJLRTXPHUGH-UHFFFAOYSA-N
2-hydroxyisonicotinic acid, 2-hydroxy-4-pyridinecarboxylic acid, 2-oxo-1,2-dihydropyridine-4-carboxylic acid, 2-hydroxypyridine-4-carboxylic acid, 2-hydroxy-isonicotinic acid, 2-hydroxyisonicotinicacid, 4-pyridinecarboxylic acid, 1,2-dihydro-2-oxo, pubchem16780, acmc-209yjf, 2-hydroxy isonicotinic acid
280756
OC(=O)C1=CC(=O)NC=C1

Used in the synthesis of selective inhibitors of neuropeptide Y Y5 inhibiting food intake. Also used in the optimization of p-subsite residues of HIV protease inhibitors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used in the synthesis of selective inhibitors of neuropeptide Y Y5 inhibiting food intake. Also used in the optimization of p-subsite residues of HIV protease inhibitors.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY
Quantity 5 g
Beilstein 115859
IUPAC Name 2-oxo-1H-pyridine-4-carboxylic acid
Formula Weight 139.11
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 2-Hydroxypyridine-4-carboxylic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'