missing translation for 'onlineSavingsMsg'
Learn More

2-Iodo-1-tritylimidazole, 97%, Thermo Scientific™

Catalog Number 15408367
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
67478-46-0
C22H17IN2
436.296
MFCD02179529
AAHIYUWXFHXGBO-UHFFFAOYSA-N
2-iodo-1-trityl-1h-imidazole, 2-iodo-1-triphenylmethyl imidazole, acmc-209nz8, 2-iodo-1-trityl-imidazole, 2-iodo-1-trityl-1h-imidazole #, imidazole, 2-iodo-1-triphenylmethyl, 2-iodo-1-triphenylmethyl-1h-imidazole, 1h-imidazole,2-iodo-1-triphenylmethyl
618240
2-iodo-1-tritylimidazole
C1=CC=C(C=C1)C(C2=CC=CC=C2)(C3=CC=CC=C3)N4C=CN=C4I
This item is not returnable. View return policy
67478-46-0
C22H17IN2
436.296
MFCD02179529
AAHIYUWXFHXGBO-UHFFFAOYSA-N
2-iodo-1-trityl-1h-imidazole, 2-iodo-1-triphenylmethyl imidazole, acmc-209nz8, 2-iodo-1-trityl-imidazole, 2-iodo-1-trityl-1h-imidazole #, imidazole, 2-iodo-1-triphenylmethyl, 2-iodo-1-triphenylmethyl-1h-imidazole, 1h-imidazole,2-iodo-1-triphenylmethyl
618240
2-iodo-1-tritylimidazole
C1=CC=C(C=C1)C(C2=CC=CC=C2)(C3=CC=CC=C3)N4C=CN=C4I

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 170°C to 171°C
Quantity 1 g
Sensitivity Light sensitive
Formula Weight 436.29
Percent Purity 97%
Chemical Name or Material 2-Iodo-1-tritylimidazole
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'