missing translation for 'onlineSavingsMsg'
Learn More

2-Iodobenzotrifluoride, 99%, Thermo Scientific™

Catalog Number p-3758238
Click to view available options
Quantity:
5 mL
25 mL
100 mL
Packaging:
Glass bottle
This item is not returnable. View return policy
444-29-1
C7H4F3I
272.01
MFCD00001041
IGZGUYVVBABKOY-UHFFFAOYSA-N
2-iodobenzotrifluoride, 1-iodo-2-trifluoromethyl benzene, o-iodobenzotrifluoride, benzene, 1-iodo-2-trifluoromethyl, 2-iodobenztrifluoride, o-trifluoromethyliodobenzene, 2-iodo-alpha,alpha,alpha-trifluorotoluene, iodobenzotrifluoride, 2-iodo-a,a,a-trifluorotoluene, 2-iodobenzotrifloride
67957
1-iodo-2-(trifluoromethyl)benzene
C1=CC=C(C(=C1)C(F)(F)F)I
This item is not returnable. View return policy
444-29-1
C7H4F3I
272.01
MFCD00001041
IGZGUYVVBABKOY-UHFFFAOYSA-N
2-iodobenzotrifluoride, 1-iodo-2-trifluoromethyl benzene, o-iodobenzotrifluoride, benzene, 1-iodo-2-trifluoromethyl, 2-iodobenztrifluoride, o-trifluoromethyliodobenzene, 2-iodo-alpha,alpha,alpha-trifluorotoluene, iodobenzotrifluoride, 2-iodo-a,a,a-trifluorotoluene, 2-iodobenzotrifloride
67957
1-iodo-2-(trifluoromethyl)benzene
C1=CC=C(C(=C1)C(F)(F)F)I
CAS Min % 98.5
CAS Max % 100.0
Color Yellow
Density 1.8900g/mL
Boiling Point 197.0°C to 198.0°C
Flash Point 81°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula CF3(C6H4)I
Refractive Index 1.5290 to 1.5310
Quantity 5 mL
Beilstein 05, III, 725
Specific Gravity 1.89
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in most common organic solvents
Formula Weight 272.01
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 2-Iodobenzotrifluoride
Show More Show Less
compliance-icons
Product Identifier
  • 2-Iodobenzotrifluoride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'