missing translation for 'onlineSavingsMsg'
Learn More

2-Mercaptoethyl sulfide, 90%, Thermo Scientific Chemicals

Catalog Number p-4440184
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
This item is not returnable. View return policy
3570-55-6
C4H10S3
154.30
MFCD00004893
KSJBMDCFYZKAFH-UHFFFAOYSA-N
2,2'-thiodiethanethiol, bis 2-mercaptoethyl sulfide, 2-mercaptoethyl sulfide, ethanethiol, 2,2'-thiobis, bis mercaptoethyl sulfide, 3-thia-1,5-pentanedithiol, 2-mercaptoethylsulfide, 2,2'-dimercaptodiethyl sulfide, unii-oeu4azc07s, ethanethiol, 2,2'-thiodi
77117
2-(2-sulfanylethylsulfanyl)ethanethiol
SCCSCCS
This item is not returnable. View return policy
3570-55-6
C4H10S3
154.30
MFCD00004893
KSJBMDCFYZKAFH-UHFFFAOYSA-N
2,2'-thiodiethanethiol, bis 2-mercaptoethyl sulfide, 2-mercaptoethyl sulfide, ethanethiol, 2,2'-thiobis, bis mercaptoethyl sulfide, 3-thia-1,5-pentanedithiol, 2-mercaptoethylsulfide, 2,2'-dimercaptodiethyl sulfide, unii-oeu4azc07s, ethanethiol, 2,2'-thiodi
77117
2-(2-sulfanylethylsulfanyl)ethanethiol
SCCSCCS

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point -11.0°C
Color Colorless
Density 1.1830g/mL
Boiling Point 135.0°C to 136.0°C (10.0 mmHg)
Flash Point 90°C
Infrared Spectrum Authentic
Assay Percent Range 88 % min. (GC)
Packaging Glass bottle
Linear Formula (HSCH2CH2)2S
Refractive Index 1.594
Quantity 10 g
Specific Gravity 1.183
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol,ether,toluene,chloroform,,dioxane,carbon tetrachloride
Formula Weight 154.32
Percent Purity 90%
Physical Form Liquid
Chemical Name or Material 2-Mercaptoethyl sulfide
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'