missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Mesitylacetonitrile, TRC
Click to view available options
Quantity:
10 g
Unit Size:
10g
Specifications
Specifications
| Chemical Name or Material | 2-Mesitylacetonitrile |
| CAS | 34688-71-6 |
| Molecular Formula | C11 H13 N |
| Quantity | 10 g |
| InChI Formula | InChI=1S/C11H13N/c1-8-6-9(2)11(4-5-12)10(3)7-8/h6-7H,4H2,1-3H3 |
| SMILES | Cc1cc(C)c(CC#N)c(C)c1 |
| IUPAC Name | 2-(2,4,6-trimethylphenyl)acetonitrile |
| Molecular Weight (g/mol) | 159.23 |
| Formula Weight | 159.1048 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction