missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxy-3-nitropyridine, 97%, Thermo Scientific™

Catalog Number p-7077516
Click to view available options
Quantity:
10 g
25 g
This item is not returnable. View return policy
20265-35-4
C6H6N2O3
154.12
WZNQCVOSOCGWJG-UHFFFAOYSA-N
2-methoxy-3-nitro-pyridine, pyridine, 2-methoxy-3-nitro, 2-methoxy-3-nitropyridin, pubchem6593, maybridge1_008636, acmc-1cdn9, 2-methoxy-3-nitropyridine?, ksc494o8j, 2-methoxy-3-nitropyridine, 2-methoxy-3-nitropyridine 1g
253219
2-methoxy-3-nitropyridine
COC1=C(C=CC=N1)[N+](=O)[O-]
This item is not returnable. View return policy
20265-35-4
C6H6N2O3
154.12
WZNQCVOSOCGWJG-UHFFFAOYSA-N
2-methoxy-3-nitro-pyridine, pyridine, 2-methoxy-3-nitro, 2-methoxy-3-nitropyridin, pubchem6593, maybridge1_008636, acmc-1cdn9, 2-methoxy-3-nitropyridine?, ksc494o8j, 2-methoxy-3-nitropyridine, 2-methoxy-3-nitropyridine 1g
253219
2-methoxy-3-nitropyridine
COC1=C(C=CC=N1)[N+](=O)[O-]
Melting Point 50.0°C to 53.0°C
Assay Percent Range 97%
Quantity 10 g
Formula Weight 154.12
Percent Purity 97%
Chemical Name or Material 2-Methoxy-3-nitropyridine
compliance-icons
Product Identifier
  • 2-Methoxy-3-nitropyridine, 97%
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'