missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Methoxyphenylacetone, TRC
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Specifications
Specifications
| Chemical Name or Material | 2-Methoxyphenylacetone |
| CAS | 5211-62-1 |
| Molecular Formula | C10 H12 O2 |
| Quantity | 100 mg |
| Synonym | 1-(2-Methoxyphenyl)-2-propanone, 2-Propanone, (o-methoxyphenyl)- (6CI,7CI), 2-Propanone, 1-(o-methoxyphenyl)- (8CI), (2-Methoxyphenyl)acetone, (o-Methoxyphenyl)acetone, 1-(2-Anisyl)-2-propanone, 1-(o-Methoxyphenyl)-2-propanone, 2'-Methoxyphenyl-2-propanone, o-Methoxyphenyl-2-propanone, 2-Methoxybenzyl methyl ketone, o-Methoxybenzyl methyl ketone, o-Methoxyphenylacetone, ortho-Methoxyphenylacetone |
| InChI Formula | InChI=1S/C10H12O2/c1-8(11)7-9-5-3-4-6-10(9)12-2/h3-6H,7H2,1-2H3 |
| SMILES | COc1ccccc1CC(=O)C |
| IUPAC Name | 1-(2-methoxyphenyl)propan-2-one |
| Molecular Weight (g/mol) | 164.2011 |
| Formula Weight | 164.0837 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction