missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Methoxyphenylacetone, TRC
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
Specifications
Specifications
| Chemical Name or Material | 2-Methoxyphenylacetone |
| CAS | 5211-62-1 |
| Molecular Formula | C10 H12 O2 |
| Quantity | 50 mg |
| Synonym | 1-(2-Methoxyphenyl)-2-propanone, 2-Propanone, (o-methoxyphenyl)- (6CI,7CI), 2-Propanone, 1-(o-methoxyphenyl)- (8CI), (2-Methoxyphenyl)acetone, (o-Methoxyphenyl)acetone, 1-(2-Anisyl)-2-propanone, 1-(o-Methoxyphenyl)-2-propanone, 2'-Methoxyphenyl-2-propanone, o-Methoxyphenyl-2-propanone, 2-Methoxybenzyl methyl ketone, o-Methoxybenzyl methyl ketone, o-Methoxyphenylacetone, ortho-Methoxyphenylacetone |
| InChI Formula | InChI=1S/C10H12O2/c1-8(11)7-9-5-3-4-6-10(9)12-2/h3-6H,7H2,1-2H3 |
| SMILES | COc1ccccc1CC(=O)C |
| IUPAC Name | 1-(2-methoxyphenyl)propan-2-one |
| Molecular Weight (g/mol) | 164.2011 |
| Formula Weight | 164.0837 |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion