missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-3-nitroaniline, 97%

Product Code. p-3758426 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 10153630

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 603-83-8 | C7H8N2O2 | 152.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 603-83-8
Molecular Formula C7H8N2O2
Molecular Weight (g/mol) 152.15
MDL Number MFCD00007731
InChI Key HFCFJYRLBAANKN-UHFFFAOYSA-N
Synonym 2-amino-6-nitrotoluene, 3-nitro-o-toluidine, benzenamine, 2-methyl-3-nitro, o-toluidine, 3-nitro, 2-methyl-3-nitro-aniline, 1-amino-2-methyl-3-nitrobenzene, 2-nitro-6-aminotoluene, 2-methyl-3-nitrobenzenamine, 2-methyl-3-nitro-phenylamine, 2-methyl-3-nitrophenyl amine
PubChem CID 11783
IUPAC Name 2-methyl-3-nitroaniline
SMILES CC1=C(N)C=CC=C1[N+]([O-])=O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 87°C to 91°C
Color Green-Yellow to Yellow
Boiling Point 305°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Linear Formula CH3C6H3(NO2)NH2
Quantity 25 g
Beilstein 12, 848
Solubility Information Solubility in water: insoluble. Other solubilities: freely soluble in ethanol and ether
Formula Weight 152.15
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 2-Methyl-3-nitroaniline
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.