missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-3-nitropyridine, 97%, Thermo Scientific™

Catalog Number p-7077429
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
18699-87-1
C6H6N2O2
138.13
MFCD04114137
CCFGTKQIRWHYTB-UHFFFAOYSA-N
2-methyl-3-nitro-pyridine, 3-nitro-2-picoline, 3-nitropicoline, 3-nitro-2-methylpyridine, pyridine, 2-methylnitro, pyridine, 2-methyl-3-nitro-9ci, nitropicoline, zlchem 137, acmc-209ep0, pyridine, 2-methyl-3-nitro
329360
2-methyl-3-nitropyridine
CC1=NC=CC=C1[N+]([O-])=O
This item is not returnable. View return policy
18699-87-1
C6H6N2O2
138.13
MFCD04114137
CCFGTKQIRWHYTB-UHFFFAOYSA-N
2-methyl-3-nitro-pyridine, 3-nitro-2-picoline, 3-nitropicoline, 3-nitro-2-methylpyridine, pyridine, 2-methylnitro, pyridine, 2-methyl-3-nitro-9ci, nitropicoline, zlchem 137, acmc-209ep0, pyridine, 2-methyl-3-nitro
329360
2-methyl-3-nitropyridine
CC1=NC=CC=C1[N+]([O-])=O
Boiling Point 104.0°C (15.0 mmHg)
Flash Point 83°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 138.13
Percent Purity 97%
Chemical Name or Material 2-Methyl-3-nitropyridine
compliance-icons
Product Identifier
  • 2-Methyl-3-nitropyridine, 97%
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'