missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-5-nitrobenzoic acid, 98+%

Product Code. p-7045755
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11342677

Brand: Thermo Scientific Alfa Aesar L11862.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

CAS: 1975-52-6 | C8H7NO4 | 181.147 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1975-52-6
Molecular Formula C8H7NO4
Molecular Weight (g/mol) 181.147
MDL Number MFCD00007371
InChI Key DJRFJAVPROZZFL-UHFFFAOYSA-N
Synonym 2-methyl-5-nitrobenzoic acid, 5-nitro-o-toluic acid, benzoic acid, 2-methyl-5-nitro, 2-methyl-5-nitro-benzoic acid, 5-nitro-2-methylbenzoic acid, rarechem al bo 0285, 5-nitro-2-methyl benzoic acid, 2-methyl-5-nitro benzoic acid, pubchem2244
PubChem CID 519683
SMILES CC1=C(C=C(C=C1)[N+](=O)[O-])C(=O)O

Specifications

Melting Point 176°C to 181°C
Quantity 1 g
Beilstein 2451300
Formula Weight 181.15
Percent Purity ≥98%
Chemical Name or Material 2-Methyl-5-nitrobenzoic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.