missing translation for 'onlineSavingsMsg'
Learn More

2-(methylamino)ethane-1-thiol hydrochloride, TRC

Product Code. 30474135
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
Product Code. Quantity unitSize
30474135 10 mg 10mg
30473242 50 mg 50mg
30464819 100 mg 100mg
3 options
This item is not returnable. View return policy

Product Code. 30474135

Brand: Toronto Research Chemicals TRCM28819010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 33744-33-1
Formula Weight 127.0222
InChI Formula InChI=1S/C3H9NS.ClH/c1-4-2-3-5;/h4-5H,2-3H2,1H3;1H
IUPAC Name 2-(methylamino)ethanethiol;hydrochloride
Molecular Formula C3H9NS.HCl
Molecular Weight (g/mol) 127.63
SMILES Cl.CNCCS
Synonym Ethanethiol, 2-(methylamino)-, hydrochloride (8CI, 9CI), 2-(Methylamino)ethane-1-thiol hydrochloride, 2-(Methylamino)ethanethiol hydrochloride, 2-Mercapto-N-methylaminoethane hydrochloride, 2-Mercapto-N-methylaminoethane hydrochloride (1:1)
Chemical Name or Material 2-(methylamino)ethane-1-thiol hydrochloride

Specifications

Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.