missing translation for 'onlineSavingsMsg'
Learn More

2-Methylbenzeneboronic acid 1,3-propanediol ester, 95%, Thermo Scientific™

Product Code. 15490056
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 15490056

Brand: Thermo Scientific Alfa Aesar H52720.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 163517-56-4
Molecular Formula C10H13BO2
Molecular Weight (g/mol) 176.022
MDL Number MFCD06659935
InChI Key UIDYFYHQHHMEKA-UHFFFAOYSA-N
Synonym 2-2-methylphenyl-1,3,2-dioxaborinane, 2-methylbenzeneboronic acid, propanediol cyclic ester, 2-methylphenylboronic acid propanediol cyclic ester, 2-o-tolyl-1,3,2-dioxaborinane, 2-2-methylphenyl 1,3,2-dioxaborinane, 1,3,2-dioxaborinane,2-2-methylphenyl, 2-methylbenzeneboronic acid 1,3-propanediol ester, 2-methylphenylboronic acid,propanediol cyclic ester, 2-methylphenylboronic acid, propanediol cyclic ester
PubChem CID 23005365
IUPAC Name 2-(2-methylphenyl)-1,3,2-dioxaborinane
SMILES B1(OCCCO1)C2=CC=CC=C2C

Specifications

Quantity 5 g
Formula Weight 176.02
Percent Purity 95%
Chemical Name or Material 2-Methylbenzeneboronic acid 1,3-propanediol ester
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.