Learn More
2-Methylimidazo[4,5-c]pyridine, 96%, Thermo Scientific Chemicals
Catalog Number p-7047406
26.86 EUR valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
250 mg
1 g
5 g
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
63604-59-1
C7H7N3
133.154
MFCD03426025
JWQVOMJORQXNME-UHFFFAOYSA-N
2-methyl-3h-imidazo 4,5-c pyridine, 2-methyl-1h-imidazo 4,5-c pyridine, 2-methylimidazo 4,5-c pyridine, 8-methyl-3-deazapurine, 8-methyl-3,7,9-triazabicyclo 4.3.0 nona-2,4,7,10-tetraene, acmc-20anq0, 2-methyl-5-azabenzimidazole, 3h-imidazo 4,5-c pyridine, 2-methyl, 1h-imidazo 4,5-c pyridine, 2-methyl
373077
2-methyl-3H-imidazo[4,5-c]pyridine
CC1=NC2=C(N1)C=NC=C2
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
63604-59-1
C7H7N3
133.154
MFCD03426025
JWQVOMJORQXNME-UHFFFAOYSA-N
2-methyl-3h-imidazo 4,5-c pyridine, 2-methyl-1h-imidazo 4,5-c pyridine, 2-methylimidazo 4,5-c pyridine, 8-methyl-3-deazapurine, 8-methyl-3,7,9-triazabicyclo 4.3.0 nona-2,4,7,10-tetraene, acmc-20anq0, 2-methyl-5-azabenzimidazole, 3h-imidazo 4,5-c pyridine, 2-methyl, 1h-imidazo 4,5-c pyridine, 2-methyl
373077
2-methyl-3H-imidazo[4,5-c]pyridine
CC1=NC2=C(N1)C=NC=C2
2-Methylimidazo[4,5-c]pyridine is used as a antagonist against platelet-activating factor (PAF).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Methylimidazo[4,5-c]pyridine is used as a antagonist against platelet-activating factor (PAF).
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 176°C to 178°C
Quantity 250 mg
Beilstein 3234
Solubility Information Insoluble in water.
Formula Weight 133.15
Percent Purity 99%
Chemical Name or Material 2-Methylimidazo[4,5-c]pyridine