Learn More
2-Methylpyrimidine-5-carboxylic acid, 97%, Thermo Scientific Chemicals
Catalog Number p-7055726
Click to view available options
Quantity:
250 mg
1 g
5 g
5194-32-1
C6H6N2O2
138.126
MFCD08235210
NMGIXZFBQPETOK-UHFFFAOYSA-N
2-methyl-5-pyrimidinecarboxylic acid, 2-methylpyrimidine-5-carboxylicacid, 5-pyrimidinecarboxylic acid, 2-methyl, 2-methyl-5-pyrimidinecarboxylicacid, pubchem13290, acmc-209kwu, ksc269i7t, 2-methyl-pyrimidine-5-carboxylic acid, 5-pyrimidinecarboxylicacid, 2-methyl, 2-methyl pyrimidine-5-carboxylic acid
13670875
2-methylpyrimidine-5-carboxylic acid
CC1=NC=C(C=N1)C(=O)O
5194-32-1
C6H6N2O2
138.126
MFCD08235210
NMGIXZFBQPETOK-UHFFFAOYSA-N
2-methyl-5-pyrimidinecarboxylic acid, 2-methylpyrimidine-5-carboxylicacid, 5-pyrimidinecarboxylic acid, 2-methyl, 2-methyl-5-pyrimidinecarboxylicacid, pubchem13290, acmc-209kwu, ksc269i7t, 2-methyl-pyrimidine-5-carboxylic acid, 5-pyrimidinecarboxylicacid, 2-methyl, 2-methyl pyrimidine-5-carboxylic acid
13670875
2-methylpyrimidine-5-carboxylic acid
CC1=NC=C(C=N1)C(=O)O
It is employed in the synthesis, spectral characterization and anticancer activity of new 2,3,6- substituted quinazolin-4(3h)-one derivatives.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is employed in the synthesis, spectral characterization and anticancer activity of new 2,3,6- substituted quinazolin-4(3h)-one derivatives.
Solubility
Slightly soluble in water.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Protect from heat. Store in freezer at -20°C.
TRUSTED_SUSTAINABILITY
Melting Point 203°C to 205°C
Quantity 250 mg
Beilstein 508773
Solubility Information Slightly soluble in water.
Formula Weight 138.13
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 2-Methylpyrimidine-5-carboxylic acid