missing translation for 'onlineSavingsMsg'
Learn More

2-n-Propylphenol, 97%, Thermo Scientific™

Product Code. 11468953
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11468953

Brand: Thermo Scientific Alfa Aesar A11088.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 644-35-9
Molecular Formula C9H12O
Molecular Weight (g/mol) 136.194
MDL Number MFCD00002251
InChI Key LCHYEKKJCUJAKN-UHFFFAOYSA-N
Synonym 2-n-propylphenol, o-propylphenol, phenol, 2-propyl, phenol, o-propyl, 1-hydroxy-2-n-propylbenzene, 1-2-hydroxyphenyl propane, propylphenol, 2-propyl-phenol, o-n-propylphenol, unii-333r6f6t3n
PubChem CID 12570
IUPAC Name 2-propylphenol
SMILES CCCC1=CC=CC=C1O

Specifications

Density 0.989
Boiling Point 214°C
Flash Point 93°C (199°F)
Odor Phenol-like
Refractive Index 1.528
Quantity 25 g
UN Number UN3145
Beilstein 1363932
Formula Weight 136.19
Percent Purity 97%
Chemical Name or Material 2-n-Propylphenol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.