missing translation for 'onlineSavingsMsg'
Learn More

2-Naphthyl acetate, 99%

Product Code. 11335189
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11335189 10 g 10g
11345189 50 g 50g
11355189 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11335189 Supplier Thermo Scientific Alfa Aesar Supplier No. A18487.09
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1523-11-1 | C12H10O2 | 186.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1523-11-1
Molecular Formula C12H10O2
Molecular Weight (g/mol) 186.21
MDL Number MFCD00004060
InChI Key RJNPPEUAJCEUPV-UHFFFAOYSA-N
Synonym 2-naphthyl acetate, 2-acetoxynaphthalene, 2-naphthalenol, acetate, beta-naphthyl acetate, 2-naphthol, acetate, naphthalene, 2-acetoxy, 2-naphthalenol, 2-acetate, .beta.-naphthol acetate, .beta.-naphthyl acetate, o-acetyl-.beta.-naphthol
PubChem CID 73709
IUPAC Name naphthalen-2-yl acetate
SMILES CC(=O)OC1=CC=C2C=CC=CC2=C1

Specifications

Melting Point 68°C to 70°C
Quantity 10 g
Beilstein 1868291
Formula Weight 186.21
Percent Purity 99%
Chemical Name or Material 2-Naphthyl acetate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.