missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Pentynoic acid, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
25 g
Description
2-Pentynoic acid is used in the stereoselective synthesis of vinylstannanes. It is also used in the synthesis of spiroindolines derivatives.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Pentynoic acid is used in the stereoselective synthesis of vinylstannanes. It is also used in the synthesis of spiroindolines derivatives.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
2-Pentynoic acid is used in the stereoselective synthesis of vinylstannanes. It is also used in the synthesis of spiroindolines derivatives.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications
Melting Point | 47°C to 53°C |
Flash Point | 96°C (205°F) |
Linear Formula | CH3CH2CH≡CCO2H |
Quantity | 25 g |
Formula Weight | 98.1 |
Percent Purity | 97% |
Chemical Name or Material | 2-Pentynoic acid |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'