missing translation for 'onlineSavingsMsg'
Learn More

2-Phenoxybenzyl bromide, 97%, Thermo Scientific Chemicals

Catalog Number p-7032207
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
82657-72-5
C13H11BrO
263.134
MFCD01320513
YQRIQBOWLXRKKG-UHFFFAOYSA-N
1-bromomethyl-2-phenoxybenzene, 2-phenoxybenzyl bromide, benzene, bromomethylphenoxy, acmc-20eqyy, 2-bromomethylphenyl phenyl ether, 1-bromomethyl-2-phenoxy-benzene, 2-bromomethyl-1-phenoxybenzene, 1-2-bromomethyl phenoxy benzene, 1-bromomethyl-2-phenyloxy benzene, benzene, 1-bromomethyl-2-phenoxy
22675469
1-(bromomethyl)-2-phenoxybenzene
C1=CC=C(C=C1)OC2=CC=CC=C2CBr
This item is not returnable. View return policy
82657-72-5
C13H11BrO
263.134
MFCD01320513
YQRIQBOWLXRKKG-UHFFFAOYSA-N
1-bromomethyl-2-phenoxybenzene, 2-phenoxybenzyl bromide, benzene, bromomethylphenoxy, acmc-20eqyy, 2-bromomethylphenyl phenyl ether, 1-bromomethyl-2-phenoxy-benzene, 2-bromomethyl-1-phenoxybenzene, 1-2-bromomethyl phenoxy benzene, 1-bromomethyl-2-phenyloxy benzene, benzene, 1-bromomethyl-2-phenoxy
22675469
1-(bromomethyl)-2-phenoxybenzene
C1=CC=C(C=C1)OC2=CC=CC=C2CBr

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
UN Number UN3265
Formula Weight 263.14
Percent Purity 97%
Chemical Name or Material 2-Phenoxybenzyl bromide

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'