missing translation for 'onlineSavingsMsg'
Learn More

2-Phenyl-1,3-thiazole-4-carboxylic acid, 97%, Thermo Scientific™

Catalog Number p-2698743
Click to view available options
Quantity:
250 mg
1 g
10 g
This item is not returnable. View return policy
7113-10-2
C10H6NO2S
204.22
MFCD00141954
IBUSLNJQKLZPNR-UHFFFAOYSA-M
2-phenylthiazole-4-carboxylic acid, 2-phenyl-4-carboxythiazole, 4-thiazolecarboxylic acid, 2-phenyl, 2-phenyl-thiazole-4-carboxylic acid, 2-phenyl-4-thiazolecarboxylic acid, 2-phenyl-1,3-thiadiazole-4-carboxylic acid, pubchem19074, 4-carboxy-2-phenylthiazole, ksc237i8r, phenyl-1,3-thiazole-4-carboxylic acid
138928
This item is not returnable. View return policy
7113-10-2
C10H6NO2S
204.22
MFCD00141954
IBUSLNJQKLZPNR-UHFFFAOYSA-M
2-phenylthiazole-4-carboxylic acid, 2-phenyl-4-carboxythiazole, 4-thiazolecarboxylic acid, 2-phenyl, 2-phenyl-thiazole-4-carboxylic acid, 2-phenyl-4-thiazolecarboxylic acid, 2-phenyl-1,3-thiadiazole-4-carboxylic acid, pubchem19074, 4-carboxy-2-phenylthiazole, ksc237i8r, phenyl-1,3-thiazole-4-carboxylic acid
138928
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 250 mg
Formula Weight 205.24
Percent Purity 97%
Chemical Name or Material 2-Phenyl-1, 3-thiazole-4-carboxylic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'