missing translation for 'onlineSavingsMsg'
Learn More

2-Phenylbutyronitrile, 95%

Product Code. 11478966
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
This item is not returnable. View return policy

Product Code. 11478966

Brand: Thermo Scientific Alfa Aesar A17093.09

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 769-68-6 | C10H11N | 145.205 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 769-68-6
Molecular Formula C10H11N
Molecular Weight (g/mol) 145.205
MDL Number MFCD00001876
InChI Key IZPUPXNVRNBDSW-UHFFFAOYSA-N
Synonym 2-phenylbutyronitrile, 2-phenylbutanentrile, 1-cyano-1-phenylpropane, butanenitrile, 2-phenyl, benzeneacetonitrile, .alpha.-ethyl, butyronitrile, 2-phenyl, benzeneacetonitrile, ?-ethyl, .alpha.-phenylbutyronitrile, alpha-phenylbutyronitrile, dl-2-phenylbutyronitrile
PubChem CID 95334
IUPAC Name 2-phenylbutanenitrile
SMILES CCC(C#N)C1=CC=CC=C1

Specifications

Density 0.975
Boiling Point 102°C to 104°C (7 mmHg)
Flash Point 105°C (221°F)
Refractive Index 1.5086
Quantity 10 g
UN Number UN3276
Formula Weight 145.2
Percent Purity 95%
Chemical Name or Material 2-Phenylbutyronitrile

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.