missing translation for 'onlineSavingsMsg'
Learn More

2-Pyridin-3-ylpiperidine, Thermo Scientific™

Catalog Number p-4510403
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
13078-04-1
C10H14N2
162.24
MFCD00006370
MTXSIJUGVMTTMU-UHFFFAOYNA-N
3-piperidin-2-yl pyridine, anabasine, dl-anabasine, neonicotine, +--anabasine, 2-pyridin-3-ylpiperidine, +/--anabasine, 3-2-piperidinyl pyridine, 2-3-pyridyl piperidine, pyridine, 3-2-piperidinyl
2181
CHEBI:28986
C1CCC(NC1)C1=CC=CN=C1
This item is not returnable. View return policy
13078-04-1
C10H14N2
162.24
MFCD00006370
MTXSIJUGVMTTMU-UHFFFAOYNA-N
3-piperidin-2-yl pyridine, anabasine, dl-anabasine, neonicotine, +--anabasine, 2-pyridin-3-ylpiperidine, +/--anabasine, 3-2-piperidinyl pyridine, 2-3-pyridyl piperidine, pyridine, 3-2-piperidinyl
2181
CHEBI:28986
C1CCC(NC1)C1=CC=CN=C1
Density 1.0460g/mL
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
IUPAC Name 3-piperidin-2-ylpyridine
Formula Weight 162.23
Percent Purity 97%
Chemical Name or Material 2-Pyridin-3-ylpiperidine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'