missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(tert-butylsulfonyl)-3-ethoxyacrylonitrile, Thermo Scientific™
Click to view available options
Quantity:
1 g
10 g
25 g
Specifications
Assay Percent Range | 97% |
Quantity | 1 g |
Formula Weight | 217.29 |
Percent Purity | 97% |
Chemical Name or Material | 2-(tert-Butylsulfonyl)-3-ethoxyacrylonitrile |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'