missing translation for 'onlineSavingsMsg'
Learn More

2-(Tetrahydropyran-4-yloxy)benzoic acid, 97%, Thermo Scientific™

Catalog Number p-3830063
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
898289-29-7
C12H14O4
222.24
MFCD09025869
AVRDARROMNUMER-UHFFFAOYSA-N
2-oxan-4-yloxy benzoic acid, 2-tetrahydropyran-4-yloxy benzoic acid, 2-oxan-4-yl oxy benzoic acid, 2-tetrahydro-2h-pyran-4-yl oxy benzoic acid, benzoic acid, 2-tetrahydro-2h-pyran-4-yl oxy, benzoic acid,2-tetrahydro-2h-pyran-4-yl oxy, 2-2h-3,4,5,6-tetrahydropyran-4-yloxy benzoic acid
21754839
2-(oxan-4-yloxy)benzoic acid
OC(=O)C1=CC=CC=C1OC1CCOCC1
This item is not returnable. View return policy
898289-29-7
C12H14O4
222.24
MFCD09025869
AVRDARROMNUMER-UHFFFAOYSA-N
2-oxan-4-yloxy benzoic acid, 2-tetrahydropyran-4-yloxy benzoic acid, 2-oxan-4-yl oxy benzoic acid, 2-tetrahydro-2h-pyran-4-yl oxy benzoic acid, benzoic acid, 2-tetrahydro-2h-pyran-4-yl oxy, benzoic acid,2-tetrahydro-2h-pyran-4-yl oxy, 2-2h-3,4,5,6-tetrahydropyran-4-yloxy benzoic acid
21754839
2-(oxan-4-yloxy)benzoic acid
OC(=O)C1=CC=CC=C1OC1CCOCC1
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 222.24
Percent Purity 97%
Chemical Name or Material 2-(Tetrahydropyran-4-yloxy)benzoic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'