missing translation for 'onlineSavingsMsg'
Learn More

2-(Trifluoromethoxy)benzoic acid, 97%, Maybridge™

Brand:  Maybridge TL00372DE

Additional Details : CAS : 1979-29-9 Poids : 0.05500kg Transport : UN number : 2811 Chem class : 6.1 Pack group : III

Quantity 5g

 View more versions of this product

Product Code. 10596785

  • 14.06€ / 5g


Expédition estimée
17-12-2020
Add to basket

Description

Description

Specifications

Chemical Identifiers

1979-29-9
206.12
JMYSPFGUBNENSE-UHFFFAOYSA-N
2777223
C1=CC=C(C(=C1)C(=O)O)OC(F)(F)F
C8H5F3O3
MFCD00052325
2-trifluoromethoxy benzoic acid, o-trifluoromethoxybenzoic acid, alpha,alpha,alpha-trifluoro-o-anisic acid, 2-trifluoromethoxy-benzoic acid, rarechem al bo 0449, pubchem4587, acmc-209f2a, ksc494o7t
2-(trifluoromethoxy)benzoic acid

Specifications

2-(Trifluoromethoxy)benzoic acid
1979-29-9
97%
CF3OC6H4CO2H
2-trifluoromethoxy benzoic acid, o-trifluoromethoxybenzoic acid, alpha,alpha,alpha-trifluoro-o-anisic acid, 2-trifluoromethoxy-benzoic acid, rarechem al bo 0449, pubchem4587, acmc-209f2a, ksc494o7t
C1=CC=C(C(=C1)C(=O)O)OC(F)(F)F
206.12
206.12
5g
97.0
C8H5F3O3
MFCD00052325
JMYSPFGUBNENSE-UHFFFAOYSA-N
2-(trifluoromethoxy)benzoic acid
2777223
97%
Videos
SDS Documents