missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-(Trifluoromethoxy)thiobenzamide, 97%

Brand:  Alfa Aesar™ H33047.06

Additional Details : CAS : 926247-77-0 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15405837

  • 240.00€ / 5g


Estimated Shipment
27-04-2021
Add to basket

Description

Description

Specifications

Chemical Identifiers

926247-77-0
221.197
WVRLRVCJDVSKRC-UHFFFAOYSA-N
16770591
C1=CC=C(C(=C1)C(=S)N)OC(F)(F)F
C8H6F3NOS
MFCD09042874
2-trifluoromethoxy thiobenzamide, 2-trifluoromethoxy benzenecarbothioamide, 2-trifluoromethoxy benzothioamide, amino 2-trifluoromethoxy phenyl methane-1-thione
2-(trifluoromethoxy)benzenecarbothioamide

Specifications

2-(Trifluoromethoxy)thiobenzamide
5g
C8H6F3NOS
2-trifluoromethoxy thiobenzamide, 2-trifluoromethoxy benzenecarbothioamide, 2-trifluoromethoxy benzothioamide, amino 2-trifluoromethoxy phenyl methane-1-thione
C1=CC=C(C(=C1)C(=S)N)OC(F)(F)F
221.197
221.2
82°C to 86°C
926247-77-0
MFCD09042874
WVRLRVCJDVSKRC-UHFFFAOYSA-N
2-(trifluoromethoxy)benzenecarbothioamide
16770591
97%
Videos
SDS Documents