missing translation for 'onlineSavingsMsg'
Learn More

2-(Trifluoromethyl)acrylic acid, 98%

Product Code. 11420597
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11420597

Brand: Thermo Scientific Alfa Aesar B22443.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 381-98-6 | C4H2F3O2 | 139.05 g/mol

2-(Trifluoromethyl)acrylic acid is used as an acidic functional monomer for molecular imprinting of nicotine. And also used in the conventional functional monomer methacrylic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(Trifluoromethyl)acrylic acid is used as an acidic functional monomer for molecular imprinting of nicotine. And also used in the conventional functional monomer methacrylic acid.

Solubility
Soluble in water.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 381-98-6
Molecular Formula C4H2F3O2
Molecular Weight (g/mol) 139.05
MDL Number MFCD00042424
InChI Key VLSRKCIBHNJFHA-UHFFFAOYSA-M
Synonym 2-trifluoromethyl acrylic acid, 2-trifluoromethyl propenoic acid, 2-trifluoromethyl prop-2-enoic acid, tfmaa, trifluoromethacrylic acid, trifluoromethylacrylic acid, 2-trifluoromethylacrylic acid, 2-trifluoromethyl-acrylic acid, 2-propenoic acid, 2-trifluoromethyl, pubchem22654
PubChem CID 587694
SMILES [O-]C(=O)C(=C)C(F)(F)F

Specifications

Melting Point 49°C to 51°C
Flash Point 82°C (180°F)
Quantity 5 g
UN Number UN3261
Solubility Information Soluble in water.
IUPAC Name 2-(trifluoromethyl)prop-2-enoate
Formula Weight 140.06
Percent Purity 98%
Chemical Name or Material 2-(Trifluoromethyl)acrylic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.